Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Species: |
Human | ||||||||||||||||||||||||||||||||||||||||||||||||||
dbSNP Submissions: |
NA
|
||||||||||||||||||||||||||||||||||||||||||||||||||
Phenotype: |
MIM: 615393 MIM: 607505 | ||||||||||||||||||||||||||||||||||||||||||||||||||
Literature Links: |
MTERF4 PubMed Links | ||||||||||||||||||||||||||||||||||||||||||||||||||
Allele Nomenclature: |
|||||||||||||||||||||||||||||||||||||||||||||||||||
Minor Allele Frequency: |
1000Genome | Applied Biosystems® | HapMap | ||||||
---|---|---|---|---|---|---|---|---|
Global
|
Caucasian - Not Available | CEPH (CEU)
|
||||||
EAS
|
African American - Not Available | YRI (Yoruba)
|
||||||
SAS
|
Chinese - Not Available | JPT (Japanese)
|
||||||
AFR
|
Japanese - Not Available | CHB (Han Chinese)
|
||||||
EUR
|
||||||||
AMR
|
MTERF4 - mitochondrial transcription termination factor 4 | ||||||
---|---|---|---|---|---|---|
There are no transcripts associated with this gene. |
PASK - PAS domain containing serine/threonine kinase | ||||||
---|---|---|---|---|---|---|
Transcript Accession | SNP Location | SNP Type | Observed Codons | Observed Amino Acid | Protein ID | |
NM_001252119.1 | Intron | NP_001239048.1 | ||||
NM_001252120.1 | Intron | NP_001239049.1 | ||||
NM_001252122.1 | Intron | NP_001239051.1 | ||||
NM_001252124.1 | Intron | NP_001239053.1 | ||||
NM_015148.3 | Intron | NP_055963.2 | ||||
XM_005246991.1 | Intron | XP_005247048.1 | ||||
XM_011510826.1 | Intron | XP_011509128.1 | ||||
XM_011510828.1 | Intron | XP_011509130.1 | ||||
XM_011510829.1 | Intron | XP_011509131.1 | ||||
XM_011510830.1 | Intron | XP_011509132.1 | ||||
XM_011510831.2 | Intron | XP_011509133.1 | ||||
XM_011510832.2 | Intron | XP_011509134.1 | ||||
XM_011510833.1 | Intron | XP_011509135.1 | ||||
XM_011510834.1 | Intron | XP_011509136.1 | ||||
XM_011510835.1 | Intron | XP_011509137.1 | ||||
XM_017003634.1 | Intron | XP_016859123.1 | ||||
XM_017003635.1 | Intron | XP_016859124.1 | ||||
XM_017003636.1 | Intron | XP_016859125.1 |
Set Membership: |
HapMap |