Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
CGATGTCACGGATAGCAGGGCAGCA[A/G]CCACCTGGCCCTGCACCTCCCTGGG
Species: |
Human | ||||||||||||||||||||||||||||||||||||||
dbSNP Submissions: |
NA
|
||||||||||||||||||||||||||||||||||||||
Phenotype: |
MIM: 602391 | ||||||||||||||||||||||||||||||||||||||
Literature Links: |
PEMT PubMed Links | ||||||||||||||||||||||||||||||||||||||
Allele Nomenclature: |
|||||||||||||||||||||||||||||||||||||||
Minor Allele Frequency: |
1000Genome | Applied Biosystems® | HapMap | |||
---|---|---|---|---|---|
Global
|
Caucasian - Not Available | CEPH (CEU) - Not Available | |||
EAS
|
African American - Not Available | YRI (Yoruba) - Not Available | |||
SAS
|
Chinese - Not Available | CHB (Han Chinese) - Not Available | |||
AFR
|
Japanese - Not Available | JPT (Japanese) - Not Available | |||
EUR
|
|||||
AMR
|
PEMT - phosphatidylethanolamine N-methyltransferase | ||||||
---|---|---|---|---|---|---|
Transcript Accession | SNP Location | SNP Type | Observed Codons | Observed Amino Acid | Protein ID | |
NM_001267551.1 | Intron | NP_001254480.1 | ||||
NM_001267552.1 | Intron | NP_001254481.1 | ||||
NM_007169.2 | Intron | NP_009100.2 | ||||
NM_148172.2 | Intron | NP_680477.1 | ||||
NM_148173.1 | Intron | NP_680478.1 | ||||
XM_006721418.3 | Intron | XP_006721481.2 | ||||
XM_017024016.1 | Intron | XP_016879505.1 |