Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
CCTAGGTTCCTTCCTGTCTCTGGGC[C/T]TCTGCTTCTTGTCTCAGCCCAGCAG
Species: |
Human | ||||||||||||||||||||||||||||||||||||||||||||||||||
dbSNP Submissions: |
44 submissions
|
||||||||||||||||||||||||||||||||||||||||||||||||||
Phenotype: |
MIM: 616793 | ||||||||||||||||||||||||||||||||||||||||||||||||||
Literature Links: |
PLA2G2F PubMed Links | ||||||||||||||||||||||||||||||||||||||||||||||||||
Allele Nomenclature: |
|||||||||||||||||||||||||||||||||||||||||||||||||||
Minor Allele Frequency: |
1000Genome | Applied Biosystems® | HapMap | ||||||
---|---|---|---|---|---|---|---|---|
Global
|
Caucasian - Not Available | CEPH (CEU)
|
||||||
EAS
|
African American - Not Available | YRI (Yoruba)
|
||||||
SAS
|
Chinese - Not Available | JPT (Japanese)
|
||||||
AFR
|
Japanese - Not Available | CHB (Han Chinese)
|
||||||
EUR
|
||||||||
AMR
|
PLA2G2F - phospholipase A2 group IIF | ||||||
---|---|---|---|---|---|---|
Transcript Accession | SNP Location | SNP Type | Observed Codons | Observed Amino Acid | Protein ID | |
NM_022819.3 | Intron | NP_073730.3 | ||||
XM_011541955.1 | Intron | XP_011540257.1 | ||||
XM_011541956.1 | Intron | XP_011540258.1 |
Set Membership: |
HapMap |