Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
TATCCAGTAGAAATTGGTGAATAGA[C/T]ATTCAATAATTTCTATGATCGTGAT
Species: |
Human | ||||||||||||||||||||||||||||||||||||||
dbSNP Submissions: |
NA
|
||||||||||||||||||||||||||||||||||||||
Phenotype: |
MIM: 616693 | ||||||||||||||||||||||||||||||||||||||
Literature Links: |
ASIC5 PubMed Links | ||||||||||||||||||||||||||||||||||||||
Allele Nomenclature: |
|||||||||||||||||||||||||||||||||||||||
Minor Allele Frequency: |
1000Genome | Applied Biosystems® | HapMap | |||
---|---|---|---|---|---|
Global
|
Caucasian - Not Available | CEPH (CEU) - Not Available | |||
EAS
|
African American - Not Available | YRI (Yoruba) - Not Available | |||
SAS
|
Chinese - Not Available | CHB (Han Chinese) - Not Available | |||
AFR
|
Japanese - Not Available | JPT (Japanese) - Not Available | |||
EUR
|
|||||
AMR
|
ASIC5 - acid sensing ion channel subunit family member 5 | ||||||
---|---|---|---|---|---|---|
Transcript Accession | SNP Location | SNP Type | Observed Codons | Observed Amino Acid | Protein ID | |
NM_017419.2 | 1444 | Missense Mutation | TAT,TGT | Y,C 466 | NP_059115.1 | |
XM_017008291.1 | 1444 | Missense Mutation | TAT,TGT | Y,C 424 | XP_016863780.1 |