Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
GGAGGCTGCGTGGCACCCTGGCTCA[A/G]CATGTTCCGGCAGCGCAGCCGCTCC
Species: |
Human | ||||||||||||||||||||
dbSNP Submissions: |
NA
|
||||||||||||||||||||
Phenotype: |
MIM: 604392 | ||||||||||||||||||||
Literature Links: |
AIPL1 PubMed Links | ||||||||||||||||||||
Allele Nomenclature: |
|||||||||||||||||||||
Minor Allele Frequency: |
1000Genome | Applied Biosystems® | HapMap |
---|---|---|
Global - Not Available | Caucasian - Not Available | CEPH (CEU) - Not Available |
EAS - Not Available | African American - Not Available | YRI (Yoruba) - Not Available |
SAS - Not Available | Chinese - Not Available | CHB (Han Chinese) - Not Available |
AFR - Not Available | Japanese - Not Available | JPT (Japanese) - Not Available |
EUR - Not Available | ||
AMR - Not Available |
AIPL1 - aryl hydrocarbon receptor interacting protein like 1 | ||||||
---|---|---|---|---|---|---|
Transcript Accession | SNP Location | SNP Type | Observed Codons | Observed Amino Acid | Protein ID | |
NM_001033054.2 | 1002 | Silent Mutation | CTG,TTG | L,L 264 | NP_001028226.1 | |
NM_001033055.2 | 1002 | Silent Mutation | CTG,TTG | L,L 267 | NP_001028227.1 | |
NM_001285399.2 | 1002 | Silent Mutation | CTG,TTG | L,L 315 | NP_001272328.1 | |
NM_001285400.2 | 1002 | Silent Mutation | CTG,TTG | L,L 305 | NP_001272329.1 | |
NM_001285401.2 | 1002 | Silent Mutation | CTG,TTG | L,L 303 | NP_001272330.1 | |
NM_001285402.1 | 1002 | Silent Mutation | CTG,TTG | L,L 288 | NP_001272331.1 | |
NM_001285403.2 | 1002 | Intron | NP_001272332.1 | |||
NM_014336.4 | 1002 | Silent Mutation | CTG,TTG | L,L 327 | NP_055151.3 |