Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
CGCGGCGCCACGCGTGTCGAGACGC[A/G]CTTCCAGCCCCGCGGCGCTGGCGAA
Species: |
Human | ||||||||||||||||||||
dbSNP Submissions: |
NA
|
||||||||||||||||||||
Phenotype: |
MIM: 613275 | ||||||||||||||||||||
Literature Links: |
FAM83D PubMed Links | ||||||||||||||||||||
Allele Nomenclature: |
|||||||||||||||||||||
Minor Allele Frequency: |
1000Genome | Applied Biosystems® | HapMap |
---|---|---|
Global - Not Available | Caucasian - Not Available | CEPH (CEU) - Not Available |
EAS - Not Available | African American - Not Available | YRI (Yoruba) - Not Available |
SAS - Not Available | Chinese - Not Available | CHB (Han Chinese) - Not Available |
AFR - Not Available | Japanese - Not Available | JPT (Japanese) - Not Available |
EUR - Not Available | ||
AMR - Not Available |
FAM83D - family with sequence similarity 83 member D | ||||||
---|---|---|---|---|---|---|
Transcript Accession | SNP Location | SNP Type | Observed Codons | Observed Amino Acid | Protein ID | |
NM_030919.2 | 532 | Missense Mutation | CAC,CGC | H,R 134 | NP_112181.3 | |
XM_017028088.1 | 532 | Intron | XP_016883577.1 |
PPP1R16B - protein phosphatase 1 regulatory subunit 16B | ||||||
---|---|---|---|---|---|---|
There are no transcripts associated with this gene. |