Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
GTCTCCCTAGCATCTCATCTCTTCC[C/G]AAACTGACCCGTCTTTTCTCTTCCC
Species: |
Human | ||||||||||||||||||||||||||||||||||||||
dbSNP Submissions: |
NA
|
||||||||||||||||||||||||||||||||||||||
Phenotype: |
MIM: 114207 MIM: 180466 | ||||||||||||||||||||||||||||||||||||||
Literature Links: |
CACNB1 PubMed Links | ||||||||||||||||||||||||||||||||||||||
Allele Nomenclature: |
|||||||||||||||||||||||||||||||||||||||
Minor Allele Frequency: |
1000Genome | Applied Biosystems® | HapMap | |||
---|---|---|---|---|---|
Global
|
Caucasian - Not Available | CEPH (CEU) - Not Available | |||
EAS
|
African American - Not Available | YRI (Yoruba) - Not Available | |||
SAS
|
Chinese - Not Available | CHB (Han Chinese) - Not Available | |||
AFR
|
Japanese - Not Available | JPT (Japanese) - Not Available | |||
EUR
|
|||||
AMR
|
CACNB1 - calcium voltage-gated channel auxiliary subunit beta 1 | ||||||
---|---|---|---|---|---|---|
There are no transcripts associated with this gene. |
RPL19 - ribosomal protein L19 | ||||||
---|---|---|---|---|---|---|
Transcript Accession | SNP Location | SNP Type | Observed Codons | Observed Amino Acid | Protein ID | |
NM_000981.3 | Intron | NP_000972.1 | ||||
XM_005257564.3 | Intron | XP_005257621.1 |
STAC2 - SH3 and cysteine rich domain 2 | ||||||
---|---|---|---|---|---|---|
There are no transcripts associated with this gene. |