Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Species: |
Human | ||||||||||||||||||||||||||||||||||||||||||||||||||
dbSNP Submissions: |
NA
|
||||||||||||||||||||||||||||||||||||||||||||||||||
Phenotype: |
MIM: 616235 | ||||||||||||||||||||||||||||||||||||||||||||||||||
Literature Links: |
KATNBL1 PubMed Links | ||||||||||||||||||||||||||||||||||||||||||||||||||
Allele Nomenclature: |
|||||||||||||||||||||||||||||||||||||||||||||||||||
Minor Allele Frequency: |
1000Genome | Applied Biosystems® | HapMap | ||||||
---|---|---|---|---|---|---|---|---|
Global
|
Caucasian - Not Available | CEPH (CEU)
|
||||||
EAS
|
African American - Not Available | YRI (Yoruba)
|
||||||
SAS
|
Chinese - Not Available | JPT (Japanese)
|
||||||
AFR
|
Japanese - Not Available | CHB (Han Chinese)
|
||||||
EUR
|
||||||||
AMR
|
KATNBL1 - katanin regulatory subunit B1 like 1 | ||||||
---|---|---|---|---|---|---|
Transcript Accession | SNP Location | SNP Type | Observed Codons | Observed Amino Acid | Protein ID | |
NM_024713.2 | Intron | NP_078989.1 | ||||
XM_005254668.3 | Intron | XP_005254725.1 | ||||
XM_017022571.1 | Intron | XP_016878060.1 | ||||
XM_017022572.1 | Intron | XP_016878061.1 | ||||
XM_017022573.1 | Intron | XP_016878062.1 | ||||
XM_017022574.1 | Intron | XP_016878063.1 | ||||
XM_017022575.1 | Intron | XP_016878064.1 |