Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
AGGGTCCGGGGGCAGTCCTAGTAGC[A/G]GCCGACCTTGGCGTCCCCAATGGCG
Species: |
Human | ||||||||||||||||||||
dbSNP Submissions: |
NA
|
||||||||||||||||||||
Phenotype: |
MIM: 605072 MIM: 601032 MIM: 176802 | ||||||||||||||||||||
Literature Links: |
GIPC1 PubMed Links | ||||||||||||||||||||
Allele Nomenclature: |
|||||||||||||||||||||
Minor Allele Frequency: |
1000Genome | Applied Biosystems® | HapMap |
---|---|---|
Global - Not Available | Caucasian - Not Available | CEPH (CEU) - Not Available |
EAS - Not Available | African American - Not Available | YRI (Yoruba) - Not Available |
SAS - Not Available | Chinese - Not Available | CHB (Han Chinese) - Not Available |
AFR - Not Available | Japanese - Not Available | JPT (Japanese) - Not Available |
EUR - Not Available | ||
AMR - Not Available |
GIPC1 - GIPC PDZ domain containing family member 1 | ||||||
---|---|---|---|---|---|---|
Transcript Accession | SNP Location | SNP Type | Observed Codons | Observed Amino Acid | Protein ID | |
NM_005716.3 | 907 | Missense Mutation | CGC,TGC | R,C 332 | NP_005707.1 | |
NM_202468.2 | 907 | Missense Mutation | CGC,TGC | R,C 332 | NP_974197.1 | |
NM_202469.2 | 907 | Missense Mutation | CGC,TGC | R,C 235 | NP_974198.1 | |
NM_202470.2 | 907 | Missense Mutation | CGC,TGC | R,C 332 | NP_974199.1 | |
NM_202494.2 | 907 | Missense Mutation | CGC,TGC | R,C 235 | NP_974223.1 | |
XM_017026147.1 | 907 | Missense Mutation | CGC,TGC | R,C 332 | XP_016881636.1 | |
XM_017026148.1 | 907 | Missense Mutation | CGC,TGC | R,C 235 | XP_016881637.1 |
PKN1 - protein kinase N1 | ||||||
---|---|---|---|---|---|---|
There are no transcripts associated with this gene. |
PTGER1 - prostaglandin E receptor 1 | ||||||
---|---|---|---|---|---|---|
There are no transcripts associated with this gene. |