Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
TAAGTATCACATTTTGAATTCTTCT[A/G]CAGAACTTACTGCCAGAAAGTCAAA
Species: |
Human | ||||||||||||||||||||||||||||||||||||||||||||||||||
dbSNP Submissions: |
NA
|
||||||||||||||||||||||||||||||||||||||||||||||||||
Phenotype: |
MIM: 608876 | ||||||||||||||||||||||||||||||||||||||||||||||||||
Literature Links: |
PCF11 PubMed Links | ||||||||||||||||||||||||||||||||||||||||||||||||||
Allele Nomenclature: |
|||||||||||||||||||||||||||||||||||||||||||||||||||
Minor Allele Frequency: |
1000Genome | Applied Biosystems® | HapMap | ||||||
---|---|---|---|---|---|---|---|---|
Global
|
Caucasian - Not Available | CEPH (CEU)
|
||||||
EAS
|
African American - Not Available | YRI (Yoruba)
|
||||||
SAS
|
Chinese - Not Available | JPT (Japanese)
|
||||||
AFR
|
Japanese - Not Available | CHB (Han Chinese)
|
||||||
EUR
|
||||||||
AMR
|
PCF11 - PCF11 cleavage and polyadenylation factor subunit | ||||||
---|---|---|---|---|---|---|
Transcript Accession | SNP Location | SNP Type | Observed Codons | Observed Amino Acid | Protein ID | |
NM_015885.3 | Intron | NP_056969.2 | ||||
XM_005274048.1 | Intron | XP_005274105.1 | ||||
XM_005274049.1 | Intron | XP_005274106.1 | ||||
XM_017017918.1 | Intron | XP_016873407.1 |
Set Membership: |
HapMap |