Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
GGCTCCTCCAGGGCCTTGCCCCGGC[A/G]GTGGTACAAGAGGACCAGCCCAGCC
Species: |
Human | ||||||||||||||||||||||||||||||||||||||||||||
dbSNP Submissions: |
NA
|
||||||||||||||||||||||||||||||||||||||||||||
Phenotype: |
MIM: 614281 MIM: 602350 MIM: 606011 | ||||||||||||||||||||||||||||||||||||||||||||
Literature Links: |
ESAM PubMed Links | ||||||||||||||||||||||||||||||||||||||||||||
Allele Nomenclature: |
|||||||||||||||||||||||||||||||||||||||||||||
Minor Allele Frequency: |
1000Genome | Applied Biosystems® | HapMap | ||||||
---|---|---|---|---|---|---|---|---|
Global
|
Caucasian - Not Available | CEPH (CEU)
|
||||||
EAS
|
African American - Not Available | YRI (Yoruba)
|
||||||
SAS
|
Chinese - Not Available | CHB (Han Chinese) - Not Available | ||||||
AFR
|
Japanese - Not Available | JPT (Japanese) - Not Available | ||||||
EUR
|
||||||||
AMR
|
ESAM - endothelial cell adhesion molecule | ||||||
---|---|---|---|---|---|---|
Transcript Accession | SNP Location | SNP Type | Observed Codons | Observed Amino Acid | Protein ID | |
NM_138961.2 | 411 | Missense Mutation | NP_620411.2 | |||
XM_005271720.4 | 411 | Missense Mutation | XP_005271777.1 | |||
XM_011543060.2 | 411 | Missense Mutation | XP_011541362.1 |
LOC101929340 - uncharacterized LOC101929340 | ||||||
---|---|---|---|---|---|---|
There are no transcripts associated with this gene. |
NRGN - neurogranin | ||||||
---|---|---|---|---|---|---|
There are no transcripts associated with this gene. |
VSIG2 - V-set and immunoglobulin domain containing 2 | ||||||
---|---|---|---|---|---|---|
There are no transcripts associated with this gene. |