Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
CGGGCCCTTTTGTTGGCCAACACCA[A/G]ACTGCGCCGGCTTGAACTGATGATT
Species: |
Human | ||||||||||||||||||||||||||||||||||||||
dbSNP Submissions: |
NA
|
||||||||||||||||||||||||||||||||||||||
Phenotype: |
MIM: 609216 | ||||||||||||||||||||||||||||||||||||||
Literature Links: |
LOC105371998 PubMed Links | ||||||||||||||||||||||||||||||||||||||
Allele Nomenclature: |
|||||||||||||||||||||||||||||||||||||||
Minor Allele Frequency: |
1000Genome | Applied Biosystems® | HapMap | |||
---|---|---|---|---|---|
Global
|
Caucasian - Not Available | CEPH (CEU) - Not Available | |||
EAS
|
African American - Not Available | YRI (Yoruba) - Not Available | |||
SAS
|
Chinese - Not Available | CHB (Han Chinese) - Not Available | |||
AFR
|
Japanese - Not Available | JPT (Japanese) - Not Available | |||
EUR
|
|||||
AMR
|
LOC105371998 - uncharacterized LOC105371998 | ||||||
---|---|---|---|---|---|---|
There are no transcripts associated with this gene. |
SPIRE1 - spire type actin nucleation factor 1 | ||||||
---|---|---|---|---|---|---|
Transcript Accession | SNP Location | SNP Type | Observed Codons | Observed Amino Acid | Protein ID | |
NM_001128626.1 | 2568 | Silent Mutation | CTG,TTG | L,L 720 | NP_001122098.1 | |
NM_001128627.1 | 2568 | Silent Mutation | CTG,TTG | L,L 586 | NP_001122099.1 | |
NM_020148.2 | 2568 | Silent Mutation | CTG,TTG | L,L 706 | NP_064533.3 | |
XM_005258122.4 | 2568 | Silent Mutation | CTG,TTG | L,L 764 | XP_005258179.1 | |
XM_011525701.1 | 2568 | Silent Mutation | CTG,TTG | L,L 778 | XP_011524003.1 | |
XM_011525702.1 | 2568 | Silent Mutation | CTG,TTG | L,L 658 | XP_011524004.1 | |
XM_011525703.2 | 2568 | Intron | XP_011524005.1 |