Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
TGTGTTTGGGGGAGTGAGAGGAATG[A/C]AGAACCACTCCCCCTATAGCCTGGC
Species: |
Human | ||||||||||||||||||||||||||||||||||||||
dbSNP Submissions: |
NA
|
||||||||||||||||||||||||||||||||||||||
Phenotype: |
MIM: 605292 | ||||||||||||||||||||||||||||||||||||||
Literature Links: |
NT5M PubMed Links | ||||||||||||||||||||||||||||||||||||||
Allele Nomenclature: |
|||||||||||||||||||||||||||||||||||||||
Minor Allele Frequency: |
1000Genome | Applied Biosystems® | HapMap | |||
---|---|---|---|---|---|
Global
|
Caucasian - Not Available | CEPH (CEU) - Not Available | |||
EAS
|
African American - Not Available | YRI (Yoruba) - Not Available | |||
SAS
|
Chinese - Not Available | CHB (Han Chinese) - Not Available | |||
AFR
|
Japanese - Not Available | JPT (Japanese) - Not Available | |||
EUR
|
|||||
AMR
|
NT5M - 5',3'-nucleotidase, mitochondrial | ||||||
---|---|---|---|---|---|---|
Transcript Accession | SNP Location | SNP Type | Observed Codons | Observed Amino Acid | Protein ID | |
NM_020201.3 | Intron | NP_064586.1 | ||||
XM_005256731.3 | Intron | XP_005256788.1 | ||||
XM_005256732.3 | Intron | XP_005256789.1 | ||||
XM_005256733.3 | Intron | XP_005256790.1 | ||||
XM_005256734.3 | Intron | XP_005256791.1 | ||||
XM_005256737.1 | Intron | XP_005256794.1 | ||||
XM_011523962.2 | Intron | XP_011522264.1 | ||||
XM_011523963.1 | Intron | XP_011522265.1 | ||||
XM_011523964.2 | Intron | XP_011522266.1 | ||||
XM_011523965.2 | Intron | XP_011522267.1 | ||||
XM_011523966.1 | Intron | XP_011522268.1 | ||||
XM_011523967.2 | Intron | XP_011522269.1 | ||||
XM_017024879.1 | Intron | XP_016880368.1 |