Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
CACTTTAACTTCATGGATGTACCTT[C/T]CTTCCTTTCCTAATGCCAACTCTGT
Species: |
Human | ||||||||||||||||||||||||||||||||||||||
dbSNP Submissions: |
NA
|
||||||||||||||||||||||||||||||||||||||
Phenotype: |
MIM: 605230 MIM: 609610 | ||||||||||||||||||||||||||||||||||||||
Literature Links: |
TP53BP1 PubMed Links | ||||||||||||||||||||||||||||||||||||||
Allele Nomenclature: |
|||||||||||||||||||||||||||||||||||||||
Minor Allele Frequency: |
1000Genome | Applied Biosystems® | HapMap | |||
---|---|---|---|---|---|
Global
|
Caucasian - Not Available | CEPH (CEU) - Not Available | |||
EAS
|
African American - Not Available | YRI (Yoruba) - Not Available | |||
SAS
|
Chinese - Not Available | CHB (Han Chinese) - Not Available | |||
AFR
|
Japanese - Not Available | JPT (Japanese) - Not Available | |||
EUR
|
|||||
AMR
|
TP53BP1 - tumor protein p53 binding protein 1 | ||||||
---|---|---|---|---|---|---|
Transcript Accession | SNP Location | SNP Type | Observed Codons | Observed Amino Acid | Protein ID | |
NM_001141979.1 | 9633 | Intron | NP_001135451.1 | |||
NM_001141980.1 | 9633 | Intron | NP_001135452.1 | |||
NM_005657.2 | 9633 | Intron | NP_005648.1 | |||
XM_011521985.1 | 9633 | UTR 3 | XP_011520287.1 | |||
XM_011521986.2 | 9633 | Intron | XP_011520288.1 | |||
XM_017022533.1 | 9633 | UTR 3 | XP_016878022.1 |
TUBGCP4 - tubulin gamma complex associated protein 4 | ||||||
---|---|---|---|---|---|---|
Transcript Accession | SNP Location | SNP Type | Observed Codons | Observed Amino Acid | Protein ID | |
NM_001286414.2 | 9633 | Intron | NP_001273343.1 | |||
NM_014444.4 | 9633 | Intron | NP_055259.2 | |||
XM_011521454.2 | 9633 | Intron | XP_011519756.1 | |||
XM_011521455.2 | 9633 | Intron | XP_011519757.1 | |||
XM_017022078.1 | 9633 | Intron | XP_016877567.1 |