Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
TCTCAAACTGGGGCACACAGCCTTT[C/T]GTGTAGATTACAATCTGCTGGTCAA
Species: |
Human | ||||||||||||||||||||||||||||||||||||||
dbSNP Submissions: |
NA
|
||||||||||||||||||||||||||||||||||||||
Phenotype: |
MIM: 613136 | ||||||||||||||||||||||||||||||||||||||
Literature Links: |
TSPAN5 PubMed Links | ||||||||||||||||||||||||||||||||||||||
Allele Nomenclature: |
|||||||||||||||||||||||||||||||||||||||
Minor Allele Frequency: |
1000Genome | Applied Biosystems® | HapMap | |||
---|---|---|---|---|---|
Global
|
Caucasian - Not Available | CEPH (CEU) - Not Available | |||
EAS
|
African American - Not Available | YRI (Yoruba) - Not Available | |||
SAS
|
Chinese - Not Available | CHB (Han Chinese) - Not Available | |||
AFR
|
Japanese - Not Available | JPT (Japanese) - Not Available | |||
EUR
|
|||||
AMR
|
TSPAN5 - tetraspanin 5 | ||||||
---|---|---|---|---|---|---|
Transcript Accession | SNP Location | SNP Type | Observed Codons | Observed Amino Acid | Protein ID | |
NM_005723.3 | 1089 | Silent Mutation | ACA,ACG | T,T 218 | NP_005714.2 | |
XM_005262680.1 | 1089 | Silent Mutation | ACA,ACG | T,T 169 | XP_005262737.1 |