Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
GTTGATGCCTCCCAGAAAAAAACGC[C/T]GCCAGCCTTCCCAGAAAGCCCCGCT
Species: |
Human | ||||||||||||||||||||||||||||||||||||||
dbSNP Submissions: |
NA
|
||||||||||||||||||||||||||||||||||||||
Phenotype: |
MIM: 602341 MIM: 614085 MIM: 604730 | ||||||||||||||||||||||||||||||||||||||
Literature Links: |
FOXM1 PubMed Links | ||||||||||||||||||||||||||||||||||||||
Allele Nomenclature: |
|||||||||||||||||||||||||||||||||||||||
Minor Allele Frequency: |
1000Genome | Applied Biosystems® | HapMap | |||
---|---|---|---|---|---|
Global
|
Caucasian - Not Available | CEPH (CEU) - Not Available | |||
EAS
|
African American - Not Available | YRI (Yoruba) - Not Available | |||
SAS
|
Chinese - Not Available | CHB (Han Chinese) - Not Available | |||
AFR
|
Japanese - Not Available | JPT (Japanese) - Not Available | |||
EUR
|
|||||
AMR
|
FOXM1 - forkhead box M1 | ||||||
---|---|---|---|---|---|---|
There are no transcripts associated with this gene. |
RHNO1 - RAD9-HUS1-RAD1 interacting nuclear orphan 1 | ||||||
---|---|---|---|---|---|---|
Transcript Accession | SNP Location | SNP Type | Observed Codons | Observed Amino Acid | Protein ID | |
NM_001252499.2 | 190 | Missense Mutation | CGC,TGC | R,C 8 | NP_001239428.1 | |
NM_001252500.2 | 190 | Missense Mutation | CGC,TGC | R,C 8 | NP_001239429.1 | |
NM_001257097.1 | 190 | Missense Mutation | CGC,TGC | R,C 8 | NP_001244026.1 | |
NM_001257098.1 | 190 | Missense Mutation | CGC,TGC | R,C 8 | NP_001244027.1 |
TULP3 - tubby like protein 3 | ||||||
---|---|---|---|---|---|---|
There are no transcripts associated with this gene. |