Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
CTGCAGCTACCTACCCGGGAGGCCC[A/G]TGCCAAGGCCTTTGGTACATGTAGT
Species: |
Human | ||||||||||||||||||||||||||||||||
dbSNP Submissions: |
NA
|
||||||||||||||||||||||||||||||||
Phenotype: |
|||||||||||||||||||||||||||||||||
Literature Links: |
C11orf42 PubMed Links | ||||||||||||||||||||||||||||||||
Allele Nomenclature: |
|||||||||||||||||||||||||||||||||
Minor Allele Frequency: |
1000Genome | Applied Biosystems® | HapMap | |||
---|---|---|---|---|---|
Global - Not Available | Caucasian - Not Available | CEPH (CEU)
|
|||
EAS - Not Available | African American - Not Available | YRI (Yoruba)
|
|||
SAS - Not Available | Chinese - Not Available | JPT (Japanese)
|
|||
AFR - Not Available | Japanese - Not Available | CHB (Han Chinese)
|
|||
EUR - Not Available | |||||
AMR - Not Available |
C11orf42 - chromosome 11 open reading frame 42 | ||||||
---|---|---|---|---|---|---|
There are no transcripts associated with this gene. |
OR52W1 - olfactory receptor family 52 subfamily W member 1 | ||||||
---|---|---|---|---|---|---|
Transcript Accession | SNP Location | SNP Type | Observed Codons | Observed Amino Acid | Protein ID | |
NM_001005178.1 | 716 | Missense Mutation | CAT,CGT | H,R 239 | NP_001005178.1 |
Set Membership: |
HapMap |