Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
GCCGCAGCCTCACAAAGACCTGGGC[C/T]TCTCGGTCACCCTTCCGGCCCTCCA
Species: |
Human | ||||||||||||||||||||||||||||||||||||||||||||||||||
dbSNP Submissions: |
NA
|
||||||||||||||||||||||||||||||||||||||||||||||||||
Phenotype: |
MIM: 608897 MIM: 616375 | ||||||||||||||||||||||||||||||||||||||||||||||||||
Literature Links: |
LOC100507191 PubMed Links | ||||||||||||||||||||||||||||||||||||||||||||||||||
Allele Nomenclature: |
|||||||||||||||||||||||||||||||||||||||||||||||||||
Minor Allele Frequency: |
1000Genome | Applied Biosystems® | HapMap | ||||||
---|---|---|---|---|---|---|---|---|
Global
|
Caucasian - Not Available | CEPH (CEU)
|
||||||
EAS
|
African American - Not Available | YRI (Yoruba)
|
||||||
SAS
|
Chinese - Not Available | JPT (Japanese)
|
||||||
AFR
|
Japanese - Not Available | CHB (Han Chinese)
|
||||||
EUR
|
||||||||
AMR
|
LOC100507191 - uncharacterized LOC100507191 | ||||||
---|---|---|---|---|---|---|
There are no transcripts associated with this gene. |
UNC13D - unc-13 homolog D | ||||||
---|---|---|---|---|---|---|
Transcript Accession | SNP Location | SNP Type | Observed Codons | Observed Amino Acid | Protein ID | |
NM_199242.2 | 3277 | Silent Mutation | GAA,GAG | E,E 1066 | NP_954712.1 | |
XM_011524504.1 | 3277 | Silent Mutation | GAA,GAG | E,E 1089 | XP_011522806.1 | |
XM_011524506.1 | 3277 | Silent Mutation | GAA,GAG | E,E 1088 | XP_011522808.1 | |
XM_011524507.2 | 3277 | Silent Mutation | GAA,GAG | E,E 886 | XP_011522809.1 |
UNK - unkempt family zinc finger | ||||||
---|---|---|---|---|---|---|
There are no transcripts associated with this gene. |