Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Species: |
Human | ||||||||||||||||||||||||||||||||||||||||||||||||||
dbSNP Submissions: |
NA
|
||||||||||||||||||||||||||||||||||||||||||||||||||
Phenotype: |
MIM: 607350 | ||||||||||||||||||||||||||||||||||||||||||||||||||
Literature Links: |
HMBOX1 PubMed Links | ||||||||||||||||||||||||||||||||||||||||||||||||||
Allele Nomenclature: |
|||||||||||||||||||||||||||||||||||||||||||||||||||
Minor Allele Frequency: |
1000Genome | Applied Biosystems® | HapMap | ||||||
---|---|---|---|---|---|---|---|---|
Global
|
Caucasian - Not Available | CEPH (CEU)
|
||||||
EAS
|
African American - Not Available | YRI (Yoruba)
|
||||||
SAS
|
Chinese - Not Available | JPT (Japanese)
|
||||||
AFR
|
Japanese - Not Available | CHB (Han Chinese)
|
||||||
EUR
|
||||||||
AMR
|
HMBOX1 - homeobox containing 1 | ||||||
---|---|---|---|---|---|---|
There are no transcripts associated with this gene. |
KIF13B - kinesin family member 13B | ||||||
---|---|---|---|---|---|---|
Transcript Accession | SNP Location | SNP Type | Observed Codons | Observed Amino Acid | Protein ID | |
NM_015254.3 | 8417 | UTR 3 | NP_056069.2 | |||
XM_005273458.1 | 8417 | UTR 3 | XP_005273515.1 | |||
XM_011544457.1 | 8417 | UTR 3 | XP_011542759.1 | |||
XM_011544458.1 | 8417 | UTR 3 | XP_011542760.1 | |||
XM_011544459.2 | 8417 | UTR 3 | XP_011542761.1 | |||
XM_011544460.2 | 8417 | UTR 3 | XP_011542762.1 | |||
XM_011544461.1 | 8417 | UTR 3 | XP_011542763.1 | |||
XM_017013257.1 | 8417 | Intron | XP_016868746.1 |
Set Membership: |
HapMap |