Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
CGCCACCTGGAAGCAGTTCTAAGGG[C/T]GAGCAGGAAGGGCAGGTGCACTCAC
Species: |
Human | ||||||||||||||||||||||||||||||||||||||
dbSNP Submissions: |
NA
|
||||||||||||||||||||||||||||||||||||||
Phenotype: |
MIM: 616787 MIM: 615648 | ||||||||||||||||||||||||||||||||||||||
Literature Links: |
CLUAP1 PubMed Links | ||||||||||||||||||||||||||||||||||||||
Allele Nomenclature: |
|||||||||||||||||||||||||||||||||||||||
Minor Allele Frequency: |
1000Genome | Applied Biosystems® | HapMap | |||
---|---|---|---|---|---|
Global
|
Caucasian - Not Available | CEPH (CEU) - Not Available | |||
EAS
|
African American - Not Available | YRI (Yoruba) - Not Available | |||
SAS
|
Chinese - Not Available | CHB (Han Chinese) - Not Available | |||
AFR
|
Japanese - Not Available | JPT (Japanese) - Not Available | |||
EUR
|
|||||
AMR
|
CLUAP1 - clusterin associated protein 1 | ||||||
---|---|---|---|---|---|---|
There are no transcripts associated with this gene. |
LOC101929732 - uncharacterized LOC101929732 | ||||||
---|---|---|---|---|---|---|
There are no transcripts associated with this gene. |
NLRC3 - NLR family CARD domain containing 3 | ||||||
---|---|---|---|---|---|---|
Transcript Accession | SNP Location | SNP Type | Observed Codons | Observed Amino Acid | Protein ID | |
NM_178844.3 | Intron | NP_849172.2 | ||||
XM_017023027.1 | Intron | XP_016878516.1 | ||||
XM_017023028.1 | Intron | XP_016878517.1 | ||||
XM_017023029.1 | Intron | XP_016878518.1 | ||||
XM_017023030.1 | Intron | XP_016878519.1 | ||||
XM_017023031.1 | Intron | XP_016878520.1 | ||||
XM_017023032.1 | Intron | XP_016878521.1 | ||||
XM_017023033.1 | Intron | XP_016878522.1 | ||||
XM_017023034.1 | Intron | XP_016878523.1 | ||||
XM_017023035.1 | Intron | XP_016878524.1 | ||||
XM_017023036.1 | Intron | XP_016878525.1 | ||||
XM_017023037.1 | Intron | XP_016878526.1 | ||||
XM_017023038.1 | Intron | XP_016878527.1 | ||||
XM_017023039.1 | Intron | XP_016878528.1 |