Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
TAGTCGGGAGTGGAGGGGCAGGATG[G/T]CACGGCCACTTGGGGCTTGGGGGCG
Species: |
Human | ||||||||||||||||||||||||||||||||||||||
dbSNP Submissions: |
NA
|
||||||||||||||||||||||||||||||||||||||
Phenotype: |
MIM: 605312 MIM: 607518 | ||||||||||||||||||||||||||||||||||||||
Literature Links: |
GDF15 PubMed Links | ||||||||||||||||||||||||||||||||||||||
Allele Nomenclature: |
|||||||||||||||||||||||||||||||||||||||
Minor Allele Frequency: |
1000Genome | Applied Biosystems® | HapMap | |||
---|---|---|---|---|---|
Global
|
Caucasian - Not Available | CEPH (CEU) - Not Available | |||
EAS
|
African American - Not Available | YRI (Yoruba) - Not Available | |||
SAS
|
Chinese - Not Available | CHB (Han Chinese) - Not Available | |||
AFR
|
Japanese - Not Available | JPT (Japanese) - Not Available | |||
EUR
|
|||||
AMR
|
GDF15 - growth differentiation factor 15 | ||||||
---|---|---|---|---|---|---|
There are no transcripts associated with this gene. |
LRRC25 - leucine rich repeat containing 25 | ||||||
---|---|---|---|---|---|---|
Transcript Accession | SNP Location | SNP Type | Observed Codons | Observed Amino Acid | Protein ID | |
NM_145256.2 | 896 | Missense Mutation | ACA,CCA | T,P 234 | NP_660299.2 | |
XM_005259739.3 | 896 | Missense Mutation | ACA,CCA | T,P 234 | XP_005259796.1 |
MIR3189 - microRNA 3189 | ||||||
---|---|---|---|---|---|---|
There are no transcripts associated with this gene. |