Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
CTCCCCCCCACCAGAGGTTAGGCTT[C/T]TTTAATCACTGCTCAGATCCATGAT
Species: |
Human | ||||||||||||||||||||||||||||||||||||||
dbSNP Submissions: |
NA
|
||||||||||||||||||||||||||||||||||||||
Phenotype: |
MIM: 612939 MIM: 610875 | ||||||||||||||||||||||||||||||||||||||
Literature Links: |
HSPBP1 PubMed Links | ||||||||||||||||||||||||||||||||||||||
Allele Nomenclature: |
|||||||||||||||||||||||||||||||||||||||
Minor Allele Frequency: |
1000Genome | Applied Biosystems® | HapMap | |||
---|---|---|---|---|---|
Global
|
Caucasian - Not Available | CEPH (CEU) - Not Available | |||
EAS
|
African American - Not Available | YRI (Yoruba) - Not Available | |||
SAS
|
Chinese - Not Available | CHB (Han Chinese) - Not Available | |||
AFR
|
Japanese - Not Available | JPT (Japanese) - Not Available | |||
EUR
|
|||||
AMR
|
HSPBP1 - HSPA (Hsp70) binding protein 1 | ||||||
---|---|---|---|---|---|---|
Transcript Accession | SNP Location | SNP Type | Observed Codons | Observed Amino Acid | Protein ID | |
NM_001130106.1 | Intron | NP_001123578.1 | ||||
NM_001297600.1 | Intron | NP_001284529.1 | ||||
NM_012267.4 | Intron | NP_036399.3 | ||||
XM_005258700.2 | Intron | XP_005258757.1 | ||||
XM_005258701.2 | Intron | XP_005258758.1 | ||||
XM_011526691.1 | Intron | XP_011524993.1 | ||||
XM_017026543.1 | Intron | XP_016882032.1 | ||||
XM_017026544.1 | Intron | XP_016882033.1 | ||||
XM_017026545.1 | Intron | XP_016882034.1 |
PPP6R1 - protein phosphatase 6 regulatory subunit 1 | ||||||
---|---|---|---|---|---|---|
There are no transcripts associated with this gene. |