Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
GAAGCCAATAATGGATATGAAAAAA[A/G]TAGAGTTTTACAGGACACATACTAG
Species: |
Human | ||||||||||||||||||||||||||||||||||||||
dbSNP Submissions: |
NA
|
||||||||||||||||||||||||||||||||||||||
Phenotype: |
|||||||||||||||||||||||||||||||||||||||
Literature Links: |
C6orf52 PubMed Links | ||||||||||||||||||||||||||||||||||||||
Allele Nomenclature: |
|||||||||||||||||||||||||||||||||||||||
Minor Allele Frequency: |
1000Genome | Applied Biosystems® | HapMap | |||
---|---|---|---|---|---|
Global
|
Caucasian - Not Available | CEPH (CEU) - Not Available | |||
EAS
|
African American - Not Available | YRI (Yoruba) - Not Available | |||
SAS
|
Chinese - Not Available | CHB (Han Chinese) - Not Available | |||
AFR
|
Japanese - Not Available | JPT (Japanese) - Not Available | |||
EUR
|
|||||
AMR
|
C6orf52 - chromosome 6 open reading frame 52 | ||||||
---|---|---|---|---|---|---|
Transcript Accession | SNP Location | SNP Type | Observed Codons | Observed Amino Acid | Protein ID | |
NM_001145020.1 | Intron | NP_001138492.1 | ||||
XM_006715088.3 | Intron | XP_006715151.1 | ||||
XM_011514572.2 | Intron | XP_011512874.1 | ||||
XM_011514574.2 | Intron | XP_011512876.1 | ||||
XM_011514575.2 | Intron | XP_011512877.1 |