Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
GAAGAGCTCCAGAGAGTGAGTCAAA[A/G]CGCTCTGTGTCCTGCTATTAATGAT
Species: |
Human | ||||||||||||||||||||||||||||||||||||||
dbSNP Submissions: |
NA
|
||||||||||||||||||||||||||||||||||||||
Phenotype: |
MIM: 614246 MIM: 614685 | ||||||||||||||||||||||||||||||||||||||
Literature Links: |
NAA60 PubMed Links | ||||||||||||||||||||||||||||||||||||||
Allele Nomenclature: |
|||||||||||||||||||||||||||||||||||||||
Minor Allele Frequency: |
1000Genome | Applied Biosystems® | HapMap | |||
---|---|---|---|---|---|
Global
|
Caucasian - Not Available | CEPH (CEU) - Not Available | |||
EAS
|
African American - Not Available | YRI (Yoruba) - Not Available | |||
SAS
|
Chinese - Not Available | CHB (Han Chinese) - Not Available | |||
AFR
|
Japanese - Not Available | JPT (Japanese) - Not Available | |||
EUR
|
|||||
AMR
|
NAA60 - N(alpha)-acetyltransferase 60, NatF catalytic subunit | ||||||
---|---|---|---|---|---|---|
Transcript Accession | SNP Location | SNP Type | Observed Codons | Observed Amino Acid | Protein ID | |
NM_001083600.2 | Intron | NP_001077069.1 | ||||
NM_001083601.2 | Intron | NP_001077070.1 | ||||
NM_001317093.1 | Intron | NP_001304022.1 | ||||
NM_001317094.1 | Intron | NP_001304023.1 | ||||
NM_001317095.1 | Intron | NP_001304024.1 | ||||
NM_001317096.1 | Intron | NP_001304025.1 | ||||
NM_001317097.1 | Intron | NP_001304026.1 | ||||
NM_001317098.1 | Intron | NP_001304027.1 | ||||
NM_024845.3 | Intron | NP_079121.1 |
ZNF597 - zinc finger protein 597 | ||||||
---|---|---|---|---|---|---|
There are no transcripts associated with this gene. |