Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
GTTTGGTTTTGAAGCTTCCCACAAA[C/T]TTAGTGTGATCTGGAGATCTAGACA
Species: |
Human | |||||||||||||||||||||||||||||||||||||||||
dbSNP Submissions: |
NA
|
|||||||||||||||||||||||||||||||||||||||||
Phenotype: |
MIM: 605584 MIM: 140210 | |||||||||||||||||||||||||||||||||||||||||
Literature Links: |
DHX38 PubMed Links | |||||||||||||||||||||||||||||||||||||||||
Allele Nomenclature: |
||||||||||||||||||||||||||||||||||||||||||
Minor Allele Frequency: |
1000Genome | Applied Biosystems® | HapMap | ||||||
---|---|---|---|---|---|---|---|---|
Global
|
Caucasian - Not Available | CEPH (CEU) - Not Available | ||||||
EAS
|
African American - Not Available | YRI (Yoruba)
|
||||||
SAS
|
Chinese - Not Available | CHB (Han Chinese) - Not Available | ||||||
AFR
|
Japanese - Not Available | JPT (Japanese) - Not Available | ||||||
EUR
|
||||||||
AMR
|
DHX38 - DEAH-box helicase 38 | ||||||
---|---|---|---|---|---|---|
There are no transcripts associated with this gene. |
HPR - haptoglobin-related protein | ||||||
---|---|---|---|---|---|---|
There are no transcripts associated with this gene. |
TXNL4B - thioredoxin like 4B | ||||||
---|---|---|---|---|---|---|
Transcript Accession | SNP Location | SNP Type | Observed Codons | Observed Amino Acid | Protein ID | |
NM_001142317.1 | 463 | Silent Mutation | AAA,AAG | K,K 101 | NP_001135789.1 | |
NM_001142318.1 | 463 | Silent Mutation | AAA,AAG | K,K 101 | NP_001135790.1 | |
NM_001324354.1 | 463 | Intron | NP_001311283.1 | |||
NM_001324355.1 | 463 | Silent Mutation | AAA,AAG | K,K 47 | NP_001311284.1 | |
NM_017853.2 | 463 | Silent Mutation | AAA,AAG | K,K 101 | NP_060323.1 | |
XM_017023377.1 | 463 | Intron | XP_016878866.1 |
Set Membership: |
HapMap |