Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
GCCCTTTTTTCGTTCCTTCCCTGTT[A/C]GGGGAAGATACACCTGGACGAGAAT
Species: |
Human | ||||||||||||||||||||||||||||||||||||||
dbSNP Submissions: |
NA
|
||||||||||||||||||||||||||||||||||||||
Phenotype: |
MIM: 605036 | ||||||||||||||||||||||||||||||||||||||
Literature Links: |
TMEM253 PubMed Links | ||||||||||||||||||||||||||||||||||||||
Allele Nomenclature: |
|||||||||||||||||||||||||||||||||||||||
Minor Allele Frequency: |
1000Genome | Applied Biosystems® | HapMap | |||
---|---|---|---|---|---|
Global
|
Caucasian - Not Available | CEPH (CEU) - Not Available | |||
EAS
|
African American - Not Available | YRI (Yoruba) - Not Available | |||
SAS
|
Chinese - Not Available | CHB (Han Chinese) - Not Available | |||
AFR
|
Japanese - Not Available | JPT (Japanese) - Not Available | |||
EUR
|
|||||
AMR
|
TMEM253 - transmembrane protein 253 | ||||||
---|---|---|---|---|---|---|
Transcript Accession | SNP Location | SNP Type | Observed Codons | Observed Amino Acid | Protein ID | |
NM_001146683.1 | 1083 | UTR 3 | NP_001140155.1 | |||
XM_006720233.3 | 1083 | UTR 3 | XP_006720296.1 | |||
XM_011537078.2 | 1083 | UTR 3 | XP_011535380.1 | |||
XM_011537079.1 | 1083 | UTR 3 | XP_011535381.1 | |||
XM_011537081.2 | 1083 | UTR 3 | XP_011535383.1 | |||
XM_011537082.2 | 1083 | UTR 3 | XP_011535384.1 | |||
XM_011537083.2 | 1083 | UTR 3 | XP_011535385.1 | |||
XM_011537084.2 | 1083 | UTR 3 | XP_011535386.1 | |||
XM_011537085.2 | 1083 | UTR 3 | XP_011535387.1 |
ZNF219 - zinc finger protein 219 | ||||||
---|---|---|---|---|---|---|
Transcript Accession | SNP Location | SNP Type | Observed Codons | Observed Amino Acid | Protein ID | |
NM_001101672.1 | 1083 | Intron | NP_001095142.1 | |||
NM_001102454.1 | 1083 | Intron | NP_001095924.1 | |||
NM_016423.2 | 1083 | Intron | NP_057507.2 | |||
XM_005267739.1 | 1083 | Intron | XP_005267796.1 | |||
XM_006720163.2 | 1083 | Intron | XP_006720226.1 | |||
XM_006720164.3 | 1083 | Intron | XP_006720227.1 | |||
XM_011536825.1 | 1083 | Intron | XP_011535127.1 | |||
XM_017021354.1 | 1083 | Intron | XP_016876843.1 | |||
XM_017021355.1 | 1083 | Intron | XP_016876844.1 |