Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Species: |
Human | ||||||||||||||||||||||||||||||||||||||||||||||||||
dbSNP Submissions: |
NA
|
||||||||||||||||||||||||||||||||||||||||||||||||||
Phenotype: |
MIM: 608918 | ||||||||||||||||||||||||||||||||||||||||||||||||||
Literature Links: |
ATPAF2 PubMed Links | ||||||||||||||||||||||||||||||||||||||||||||||||||
Allele Nomenclature: |
|||||||||||||||||||||||||||||||||||||||||||||||||||
Minor Allele Frequency: |
1000Genome | Applied Biosystems® | HapMap | ||||||
---|---|---|---|---|---|---|---|---|
Global
|
Caucasian - Not Available | CEPH (CEU)
|
||||||
EAS
|
African American - Not Available | YRI (Yoruba)
|
||||||
SAS
|
Chinese - Not Available | JPT (Japanese)
|
||||||
AFR
|
Japanese - Not Available | CHB (Han Chinese)
|
||||||
EUR
|
||||||||
AMR
|
ATPAF2 - ATP synthase mitochondrial F1 complex assembly factor 2 | ||||||
---|---|---|---|---|---|---|
Transcript Accession | SNP Location | SNP Type | Observed Codons | Observed Amino Acid | Protein ID | |
NM_145691.3 | Intron | NP_663729.1 | ||||
XM_005256848.3 | Intron | XP_005256905.1 | ||||
XM_011524065.1 | Intron | XP_011522367.1 | ||||
XM_017025302.1 | Intron | XP_016880791.1 | ||||
XM_017025303.1 | Intron | XP_016880792.1 |
DRC3 - dynein regulatory complex subunit 3 | ||||||
---|---|---|---|---|---|---|
Transcript Accession | SNP Location | SNP Type | Observed Codons | Observed Amino Acid | Protein ID | |
NM_001130090.1 | Intron | NP_001123562.1 | ||||
NM_001130091.1 | Intron | NP_001123563.1 | ||||
NM_001130092.1 | Intron | NP_001123564.1 | ||||
NM_031294.3 | Intron | NP_112584.3 | ||||
XM_006721583.3 | Intron | XP_006721646.1 | ||||
XM_011524018.2 | Intron | XP_011522320.1 | ||||
XM_011524019.2 | Intron | XP_011522321.1 | ||||
XM_011524020.2 | Intron | XP_011522322.1 | ||||
XM_011524021.2 | Intron | XP_011522323.1 | ||||
XM_011524022.2 | Intron | XP_011522324.1 | ||||
XM_011524023.2 | Intron | XP_011522325.1 | ||||
XM_011524025.2 | Intron | XP_011522327.1 | ||||
XM_011524026.2 | Intron | XP_011522328.1 |
Set Membership: |
HapMap |