Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
GGCAGAGCCGGACTCGGTGCATTTC[C/T]GTGTCTTTAGAGGAGAAGGCTTTTT
Species: |
Human | ||||||||||||||||||||||||||||||||||||||
dbSNP Submissions: |
NA
|
||||||||||||||||||||||||||||||||||||||
Phenotype: |
|||||||||||||||||||||||||||||||||||||||
Literature Links: |
COQ10B PubMed Links | ||||||||||||||||||||||||||||||||||||||
Allele Nomenclature: |
|||||||||||||||||||||||||||||||||||||||
Minor Allele Frequency: |
1000Genome | Applied Biosystems® | HapMap | |||
---|---|---|---|---|---|
Global
|
Caucasian - Not Available | CEPH (CEU) - Not Available | |||
EAS
|
African American - Not Available | YRI (Yoruba) - Not Available | |||
SAS
|
Chinese - Not Available | CHB (Han Chinese) - Not Available | |||
AFR
|
Japanese - Not Available | JPT (Japanese) - Not Available | |||
EUR
|
|||||
AMR
|
COQ10B - coenzyme Q10B | ||||||
---|---|---|---|---|---|---|
Transcript Accession | SNP Location | SNP Type | Observed Codons | Observed Amino Acid | Protein ID | |
NM_001320818.1 | 10 | Intron | NP_001307747.1 | |||
NM_001320819.1 | 10 | UTR 5 | NP_001307748.1 | |||
NM_001320820.1 | 10 | UTR 5 | NP_001307749.1 | |||
NM_025147.4 | 10 | Intron | NP_079423.1 |