Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
ACATTCACAGGCATTCTGTGCCATA[C/T]GCAAATTGTTAGGTACAGGAAACAT
Species: |
Human | ||||||||||||||||||||
dbSNP Submissions: |
NA
|
||||||||||||||||||||
Phenotype: |
MIM: 194538 | ||||||||||||||||||||
Literature Links: |
ZNF10 PubMed Links | ||||||||||||||||||||
Allele Nomenclature: |
|||||||||||||||||||||
Minor Allele Frequency: |
1000Genome | Applied Biosystems® | HapMap |
---|---|---|
Global - Not Available | Caucasian - Not Available | CEPH (CEU) - Not Available |
EAS - Not Available | African American - Not Available | YRI (Yoruba) - Not Available |
SAS - Not Available | Chinese - Not Available | CHB (Han Chinese) - Not Available |
AFR - Not Available | Japanese - Not Available | JPT (Japanese) - Not Available |
EUR - Not Available | ||
AMR - Not Available |
ZNF10 - zinc finger protein 10 | ||||||
---|---|---|---|---|---|---|
There are no transcripts associated with this gene. |
ZNF891 - zinc finger protein 891 | ||||||
---|---|---|---|---|---|---|
Transcript Accession | SNP Location | SNP Type | Observed Codons | Observed Amino Acid | Protein ID | |
NM_001277291.1 | 1054 | Missense Mutation | CAT,CGT | H,R 288 | NP_001264220.1 | |
XM_017018666.1 | 1054 | Missense Mutation | CAT,CGT | H,R 288 | XP_016874155.1 | |
XM_017018667.1 | 1054 | Missense Mutation | CAT,CGT | H,R 288 | XP_016874156.1 | |
XM_017018668.1 | 1054 | Missense Mutation | CAT,CGT | H,R 288 | XP_016874157.1 |