Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
TGAAGTGCTTAATTACTTTGTTTAT[A/G]TTCCTCCCTTTAGCAGATTAACATT
Species: |
Human | ||||||||||||||||||||||||||||||||||||||||||||||||||
dbSNP Submissions: |
NA
|
||||||||||||||||||||||||||||||||||||||||||||||||||
Phenotype: |
|||||||||||||||||||||||||||||||||||||||||||||||||||
Literature Links: |
CCDC173 PubMed Links | ||||||||||||||||||||||||||||||||||||||||||||||||||
Allele Nomenclature: |
|||||||||||||||||||||||||||||||||||||||||||||||||||
Minor Allele Frequency: |
1000Genome | Applied Biosystems® | HapMap | ||||||
---|---|---|---|---|---|---|---|---|
Global
|
Caucasian - Not Available | CEPH (CEU)
|
||||||
EAS
|
African American - Not Available | YRI (Yoruba)
|
||||||
SAS
|
Chinese - Not Available | JPT (Japanese)
|
||||||
AFR
|
Japanese - Not Available | CHB (Han Chinese)
|
||||||
EUR
|
||||||||
AMR
|
CCDC173 - coiled-coil domain containing 173 | ||||||
---|---|---|---|---|---|---|
There are no transcripts associated with this gene. |
PHOSPHO2 - phosphatase, orphan 2 | ||||||
---|---|---|---|---|---|---|
Transcript Accession | SNP Location | SNP Type | Observed Codons | Observed Amino Acid | Protein ID | |
NM_001008489.3 | Intron | NP_001008489.1 | ||||
NM_001199285.1 | Intron | NP_001186214.1 | ||||
NM_001199286.1 | Intron | NP_001186215.1 | ||||
NM_001199287.1 | Intron | NP_001186216.1 | ||||
NM_001199288.1 | Intron | NP_001186217.1 |
PHOSPHO2-KLHL23 - PHOSPHO2-KLHL23 readthrough | ||||||
---|---|---|---|---|---|---|
Transcript Accession | SNP Location | SNP Type | Observed Codons | Observed Amino Acid | Protein ID | |
NM_001199290.1 | Intron | NP_001186219.1 |
Set Membership: |
HapMap |