Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
CTGTGGTATACTGATGAGCAGAACT[C/T]TTCCACTACATACCTCAATTTTGCC
Species: |
Human | ||||||||||||||||||||
dbSNP Submissions: |
NA
|
||||||||||||||||||||
Phenotype: |
|||||||||||||||||||||
Literature Links: |
COQ10B PubMed Links | ||||||||||||||||||||
Allele Nomenclature: |
|||||||||||||||||||||
Minor Allele Frequency: |
1000Genome | Applied Biosystems® | HapMap |
---|---|---|
Global - Not Available | Caucasian - Not Available | CEPH (CEU) - Not Available |
EAS - Not Available | African American - Not Available | YRI (Yoruba) - Not Available |
SAS - Not Available | Chinese - Not Available | CHB (Han Chinese) - Not Available |
AFR - Not Available | Japanese - Not Available | JPT (Japanese) - Not Available |
EUR - Not Available | ||
AMR - Not Available |
COQ10B - coenzyme Q10B | ||||||
---|---|---|---|---|---|---|
Transcript Accession | SNP Location | SNP Type | Observed Codons | Observed Amino Acid | Protein ID | |
NM_001320818.1 | 324 | Intron | NP_001307747.1 | |||
NM_001320819.1 | 324 | Missense Mutation | CTT,TTT | L,F 20 | NP_001307748.1 | |
NM_001320820.1 | 324 | Missense Mutation | CTT,TTT | L,F 5 | NP_001307749.1 | |
NM_025147.4 | 324 | Missense Mutation | CTT,TTT | L,F 48 | NP_079423.1 |