Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
CATCTGAATCAGAGACAGGGAAGAA[C/T]AGGCTGAGGCCTGCTGCGGAGGAGG
Species: |
Human | ||||||||||||||||||||||||||||||||||||||
dbSNP Submissions: |
NA
|
||||||||||||||||||||||||||||||||||||||
Phenotype: |
MIM: 610503 MIM: 141251 | ||||||||||||||||||||||||||||||||||||||
Literature Links: |
CDIP1 PubMed Links | ||||||||||||||||||||||||||||||||||||||
Allele Nomenclature: |
|||||||||||||||||||||||||||||||||||||||
Minor Allele Frequency: |
1000Genome | Applied Biosystems® | HapMap | |||
---|---|---|---|---|---|
Global
|
Caucasian - Not Available | CEPH (CEU) - Not Available | |||
EAS
|
African American - Not Available | YRI (Yoruba) - Not Available | |||
SAS
|
Chinese - Not Available | CHB (Han Chinese) - Not Available | |||
AFR
|
Japanese - Not Available | JPT (Japanese) - Not Available | |||
EUR
|
|||||
AMR
|
CDIP1 - cell death-inducing p53 target 1 | ||||||
---|---|---|---|---|---|---|
Transcript Accession | SNP Location | SNP Type | Observed Codons | Observed Amino Acid | Protein ID | |
NM_001199054.1 | Intron | NP_001185983.1 | ||||
NM_001199055.1 | Intron | NP_001185984.1 | ||||
NM_001199056.1 | Intron | NP_001185985.1 | ||||
NM_013399.2 | Intron | NP_037531.2 |
HMOX2 - heme oxygenase 2 | ||||||
---|---|---|---|---|---|---|
There are no transcripts associated with this gene. |