Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
CCAGGGTCGCTGAGACAGGAGGGCC[A/G]GGACGGGACCATGGAGAGTGGCCAA
Species: |
Human | ||||||||||||||||||||||||||||||||||||||
dbSNP Submissions: |
NA
|
||||||||||||||||||||||||||||||||||||||
Phenotype: |
MIM: 608897 MIM: 616375 | ||||||||||||||||||||||||||||||||||||||
Literature Links: |
LOC100507191 PubMed Links | ||||||||||||||||||||||||||||||||||||||
Allele Nomenclature: |
|||||||||||||||||||||||||||||||||||||||
Minor Allele Frequency: |
1000Genome | Applied Biosystems® | HapMap | |||
---|---|---|---|---|---|
Global
|
Caucasian - Not Available | CEPH (CEU) - Not Available | |||
EAS
|
African American - Not Available | YRI (Yoruba) - Not Available | |||
SAS
|
Chinese - Not Available | CHB (Han Chinese) - Not Available | |||
AFR
|
Japanese - Not Available | JPT (Japanese) - Not Available | |||
EUR
|
|||||
AMR
|
LOC100507191 - uncharacterized LOC100507191 | ||||||
---|---|---|---|---|---|---|
There are no transcripts associated with this gene. |
UNC13D - unc-13 homolog D | ||||||
---|---|---|---|---|---|---|
Transcript Accession | SNP Location | SNP Type | Observed Codons | Observed Amino Acid | Protein ID | |
NM_199242.2 | Intron | NP_954712.1 | ||||
XM_011524504.1 | Intron | XP_011522806.1 | ||||
XM_011524506.1 | Intron | XP_011522808.1 | ||||
XM_011524507.2 | Intron | XP_011522809.1 |
UNK - unkempt family zinc finger | ||||||
---|---|---|---|---|---|---|
There are no transcripts associated with this gene. |