Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
GGTGTGAATCTCCCACTTCCTGAGG[G/T]GGCATTTAGTGGAGAACAAGAATTA
Species: |
Human | |||||||||||||||||||||||
dbSNP Submissions: |
NA
|
|||||||||||||||||||||||
Phenotype: |
MIM: 613447 | |||||||||||||||||||||||
Literature Links: |
CFAP44 PubMed Links | |||||||||||||||||||||||
Allele Nomenclature: |
||||||||||||||||||||||||
Minor Allele Frequency: |
1000Genome | Applied Biosystems® | HapMap | |||
---|---|---|---|---|---|
Global
|
Caucasian - Not Available | CEPH (CEU) - Not Available | |||
EAS - Not Available | African American - Not Available | YRI (Yoruba) - Not Available | |||
SAS - Not Available | Chinese - Not Available | CHB (Han Chinese) - Not Available | |||
AFR - Not Available | Japanese - Not Available | JPT (Japanese) - Not Available | |||
EUR - Not Available | |||||
AMR - Not Available |
CFAP44 - cilia and flagella associated protein 44 | ||||||
---|---|---|---|---|---|---|
There are no transcripts associated with this gene. |
SPICE1 - spindle and centriole associated protein 1 | ||||||
---|---|---|---|---|---|---|
Transcript Accession | SNP Location | SNP Type | Observed Codons | Observed Amino Acid | Protein ID | |
NM_144718.3 | 2655 | Missense Mutation | ACC,CCC | T,P 824 | NP_653319.1 | |
XM_011512461.2 | 2655 | Missense Mutation | ACC,CCC | T,P 838 | XP_011510763.1 | |
XM_017005775.1 | 2655 | Missense Mutation | ACC,CCC | T,P 838 | XP_016861264.1 | |
XM_017005776.1 | 2655 | Missense Mutation | ACC,CCC | T,P 836 | XP_016861265.1 | |
XM_017005777.1 | 2655 | Missense Mutation | ACC,CCC | T,P 824 | XP_016861266.1 | |
XM_017005778.1 | 2655 | Missense Mutation | ACC,CCC | T,P 824 | XP_016861267.1 |