Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
TGTCCAGCGCCAGGAGCCCTCTGTG[C/T]CCCTGTCAGTGAGACCGATCCAGTA
Species: |
Human | ||||||||||||||||||||||||||||||||||||||
dbSNP Submissions: |
NA
|
||||||||||||||||||||||||||||||||||||||
Phenotype: |
|||||||||||||||||||||||||||||||||||||||
Literature Links: |
CLEC4F PubMed Links | ||||||||||||||||||||||||||||||||||||||
Allele Nomenclature: |
|||||||||||||||||||||||||||||||||||||||
Minor Allele Frequency: |
1000Genome | Applied Biosystems® | HapMap | |||
---|---|---|---|---|---|
Global
|
Caucasian - Not Available | CEPH (CEU) - Not Available | |||
EAS
|
African American - Not Available | YRI (Yoruba) - Not Available | |||
SAS
|
Chinese - Not Available | CHB (Han Chinese) - Not Available | |||
AFR
|
Japanese - Not Available | JPT (Japanese) - Not Available | |||
EUR
|
|||||
AMR
|
CLEC4F - C-type lectin domain family 4 member F | ||||||
---|---|---|---|---|---|---|
Transcript Accession | SNP Location | SNP Type | Observed Codons | Observed Amino Acid | Protein ID | |
NM_001258027.1 | 1675 | Missense Mutation | GAC,GGC | D,G 533 | NP_001244956.1 | |
NM_001321308.1 | 1675 | Missense Mutation | GAC,GGC | D,G 533 | NP_001308237.1 | |
NM_173535.2 | 1675 | Missense Mutation | GAC,GGC | D,G 533 | NP_775806.2 | |
XM_011532635.2 | 1675 | Missense Mutation | GAC,GGC | D,G 643 | XP_011530937.1 | |
XM_011532637.2 | 1675 | Missense Mutation | GAC,GGC | D,G 643 | XP_011530939.1 | |
XM_011532638.2 | 1675 | Missense Mutation | GAC,GGC | D,G 643 | XP_011530940.1 | |
XM_011532639.2 | 1675 | Missense Mutation | GAC,GGC | D,G 643 | XP_011530941.1 | |
XM_011532640.2 | 1675 | Missense Mutation | GAC,GGC | D,G 643 | XP_011530942.1 | |
XM_011532641.2 | 1675 | UTR 3 | XP_011530943.1 | |||
XM_011532642.2 | 1675 | Missense Mutation | GAC,GGC | D,G 503 | XP_011530944.1 | |
XM_011532643.1 | 1675 | Missense Mutation | GAC,GGC | D,G 472 | XP_011530945.1 | |
XM_017003519.1 | 1675 | Missense Mutation | GAC,GGC | D,G 643 | XP_016859008.1 |