Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
TGACCCTCATGGACAGAGTGTGAGC[A/C]AGGAGGATTTTCTCAAAGGCAGCGG
Species: |
Human | ||||||||||||||||||||||||||||||||||||||
dbSNP Submissions: |
NA
|
||||||||||||||||||||||||||||||||||||||
Phenotype: |
MIM: 613452 | ||||||||||||||||||||||||||||||||||||||
Literature Links: |
CATSPERG PubMed Links | ||||||||||||||||||||||||||||||||||||||
Allele Nomenclature: |
|||||||||||||||||||||||||||||||||||||||
Minor Allele Frequency: |
1000Genome | Applied Biosystems® | HapMap | |||
---|---|---|---|---|---|
Global
|
Caucasian - Not Available | CEPH (CEU) - Not Available | |||
EAS
|
African American - Not Available | YRI (Yoruba) - Not Available | |||
SAS
|
Chinese - Not Available | CHB (Han Chinese) - Not Available | |||
AFR
|
Japanese - Not Available | JPT (Japanese) - Not Available | |||
EUR
|
|||||
AMR
|
CATSPERG - cation channel sperm associated auxiliary subunit gamma | ||||||
---|---|---|---|---|---|---|
Transcript Accession | SNP Location | SNP Type | Observed Codons | Observed Amino Acid | Protein ID | |
NM_021185.4 | Intron | NP_067008.3 | ||||
XM_005259114.4 | Intron | XP_005259171.1 | ||||
XM_005259115.4 | Intron | XP_005259172.1 | ||||
XM_005259116.2 | Intron | XP_005259173.1 | ||||
XM_006723308.3 | Intron | XP_006723371.1 | ||||
XM_006723310.2 | Intron | XP_006723373.1 | ||||
XM_006723311.2 | Intron | XP_006723374.1 | ||||
XM_011527174.2 | Intron | XP_011525476.1 | ||||
XM_011527175.2 | Intron | XP_011525477.1 | ||||
XM_011527176.2 | Intron | XP_011525478.1 | ||||
XM_011527177.2 | Intron | XP_011525479.1 | ||||
XM_011527178.2 | Intron | XP_011525480.1 | ||||
XM_011527179.1 | Intron | XP_011525481.1 | ||||
XM_011527182.2 | Intron | XP_011525484.1 | ||||
XM_017027053.1 | Intron | XP_016882542.1 | ||||
XM_017027054.1 | Intron | XP_016882543.1 | ||||
XM_017027055.1 | Intron | XP_016882544.1 |