Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Species: |
Human | ||||||||||||||||||||||||||||||||||||||
dbSNP Submissions: |
NA
|
||||||||||||||||||||||||||||||||||||||
Phenotype: |
MIM: 602446 | ||||||||||||||||||||||||||||||||||||||
Literature Links: |
GPC5 PubMed Links | ||||||||||||||||||||||||||||||||||||||
Allele Nomenclature: |
|||||||||||||||||||||||||||||||||||||||
Minor Allele Frequency: |
1000Genome | Applied Biosystems® | HapMap | |||
---|---|---|---|---|---|
Global
|
Caucasian - Not Available | CEPH (CEU) - Not Available | |||
EAS
|
African American - Not Available | YRI (Yoruba) - Not Available | |||
SAS
|
Chinese - Not Available | CHB (Han Chinese) - Not Available | |||
AFR
|
Japanese - Not Available | JPT (Japanese) - Not Available | |||
EUR
|
|||||
AMR
|
GPC5 - glypican 5 | ||||||
---|---|---|---|---|---|---|
Transcript Accession | SNP Location | SNP Type | Observed Codons | Observed Amino Acid | Protein ID | |
NM_004466.5 | Intron | NP_004457.1 | ||||
XM_011521054.2 | Intron | XP_011519356.1 | ||||
XM_011521055.2 | Intron | XP_011519357.1 | ||||
XM_011521056.2 | Intron | XP_011519358.1 | ||||
XM_011521057.2 | Intron | XP_011519359.1 | ||||
XM_011521058.2 | Intron | XP_011519360.1 | ||||
XM_011521059.2 | Intron | XP_011519361.1 | ||||
XM_011521060.2 | Intron | XP_011519362.1 | ||||
XM_017020435.1 | Intron | XP_016875924.1 | ||||
XM_017020436.1 | Intron | XP_016875925.1 | ||||
XM_017020437.1 | Intron | XP_016875926.1 |