Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
TTTTCATGTTCCTACCAATTTTGAA[G/A]AAATAGTTTCATCTTAGCAGATGAA
Species: |
Human | ||||||||||||||||||||||||||||||||||||||
dbSNP Submissions: |
NA
|
||||||||||||||||||||||||||||||||||||||
Phenotype: |
MIM: 616643 MIM: 606562 | ||||||||||||||||||||||||||||||||||||||
Literature Links: |
HDGFRP3 PubMed Links | ||||||||||||||||||||||||||||||||||||||
Allele Nomenclature: |
|||||||||||||||||||||||||||||||||||||||
Minor Allele Frequency: |
1000Genome | Applied Biosystems® | HapMap | |||
---|---|---|---|---|---|
Global
|
Caucasian - Not Available | CEPH (CEU) - Not Available | |||
EAS
|
African American - Not Available | YRI (Yoruba) - Not Available | |||
SAS
|
Chinese - Not Available | CHB (Han Chinese) - Not Available | |||
AFR
|
Japanese - Not Available | JPT (Japanese) - Not Available | |||
EUR
|
|||||
AMR
|
HDGFRP3 - hepatoma-derived growth factor, related protein 3 | ||||||
---|---|---|---|---|---|---|
Transcript Accession | SNP Location | SNP Type | Observed Codons | Observed Amino Acid | Protein ID | |
NM_016073.3 | Intron | NP_057157.1 | ||||
XM_006720554.3 | Intron | XP_006720617.1 |
LOC105370931 - uncharacterized LOC105370931 | ||||||
---|---|---|---|---|---|---|
There are no transcripts associated with this gene. |
TM6SF1 - transmembrane 6 superfamily member 1 | ||||||
---|---|---|---|---|---|---|
Transcript Accession | SNP Location | SNP Type | Observed Codons | Observed Amino Acid | Protein ID | |
NM_001144903.1 | Intron | NP_001138375.1 | ||||
NM_023003.3 | Intron | NP_075379.2 | ||||
XM_011521679.2 | Intron | XP_011519981.1 | ||||
XM_011521680.2 | Intron | XP_011519982.1 | ||||
XM_017022320.1 | Intron | XP_016877809.1 | ||||
XM_017022321.1 | Intron | XP_016877810.1 | ||||
XM_017022322.1 | Intron | XP_016877811.1 | ||||
XM_017022323.1 | Intron | XP_016877812.1 | ||||
XM_017022324.1 | Intron | XP_016877813.1 | ||||
XM_017022325.1 | Intron | XP_016877814.1 | ||||
XM_017022326.1 | Intron | XP_016877815.1 | ||||
XM_017022327.1 | Intron | XP_016877816.1 | ||||
XM_017022328.1 | Intron | XP_016877817.1 | ||||
XM_017022329.1 | Intron | XP_016877818.1 | ||||
XM_017022330.1 | Intron | XP_016877819.1 | ||||
XM_017022331.1 | Intron | XP_016877820.1 |