Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
ATTATTATATATGTGTGAAATTCCA[C/T]TATTAATAGTATAGCTCCCAGCTGA
Species: |
Human | ||||||||||||||||||||||||||||||||||||||
dbSNP Submissions: |
NA
|
||||||||||||||||||||||||||||||||||||||
Phenotype: |
MIM: 605073 | ||||||||||||||||||||||||||||||||||||||
Literature Links: |
PPM1E PubMed Links | ||||||||||||||||||||||||||||||||||||||
Allele Nomenclature: |
|||||||||||||||||||||||||||||||||||||||
Minor Allele Frequency: |
1000Genome | Applied Biosystems® | HapMap | |||
---|---|---|---|---|---|
Global
|
Caucasian - Not Available | CEPH (CEU) - Not Available | |||
EAS
|
African American - Not Available | YRI (Yoruba) - Not Available | |||
SAS
|
Chinese - Not Available | CHB (Han Chinese) - Not Available | |||
AFR
|
Japanese - Not Available | JPT (Japanese) - Not Available | |||
EUR
|
|||||
AMR
|
PPM1E - protein phosphatase, Mg2+/Mn2+ dependent 1E | ||||||
---|---|---|---|---|---|---|
Transcript Accession | SNP Location | SNP Type | Observed Codons | Observed Amino Acid | Protein ID | |
NM_014906.4 | Intron | NP_055721.3 | ||||
XM_005257169.4 | Intron | XP_005257226.1 | ||||
XM_011524534.2 | Intron | XP_011522836.1 | ||||
XM_017024359.1 | Intron | XP_016879848.1 |