Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
ATAGTCGATACCCCCTGTTTCCAAA[C/G]ATAGTGGTAACTCTTAAGAAAATAA
Species: |
Human | ||||||||||||||||||||||||||||||||||||||
dbSNP Submissions: |
NA
|
||||||||||||||||||||||||||||||||||||||
Phenotype: |
MIM: 300684 | ||||||||||||||||||||||||||||||||||||||
Literature Links: |
XKRX PubMed Links | ||||||||||||||||||||||||||||||||||||||
Allele Nomenclature: |
|||||||||||||||||||||||||||||||||||||||
Minor Allele Frequency: |
1000Genome | Applied Biosystems® | HapMap | |||
---|---|---|---|---|---|
Global
|
Caucasian - Not Available | CEPH (CEU) - Not Available | |||
EAS
|
African American - Not Available | YRI (Yoruba) - Not Available | |||
SAS
|
Chinese - Not Available | CHB (Han Chinese) - Not Available | |||
AFR
|
Japanese - Not Available | JPT (Japanese) - Not Available | |||
EUR
|
|||||
AMR
|
XKRX - XK related, X-linked | ||||||
---|---|---|---|---|---|---|
Transcript Accession | SNP Location | SNP Type | Observed Codons | Observed Amino Acid | Protein ID | |
NM_212559.2 | 1506 | UTR 3 | NP_997724.2 | |||
XM_005262130.2 | 1506 | UTR 3 | XP_005262187.1 | |||
XM_011530954.2 | 1506 | Intron | XP_011529256.1 | |||
XM_011530955.1 | 1506 | UTR 3 | XP_011529257.1 | |||
XM_017029517.1 | 1506 | UTR 3 | XP_016885006.1 |