Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
TCCTTCAGCAAGCTGTTGACAGAGC[C/T]CAGCAGCTCCTGGAAGTAGCCCTCG
Species: |
Human | ||||||||||||||||||||||||||||||||||||||
dbSNP Submissions: |
NA
|
||||||||||||||||||||||||||||||||||||||
Phenotype: |
MIM: 608005 | ||||||||||||||||||||||||||||||||||||||
Literature Links: |
SIL1 PubMed Links | ||||||||||||||||||||||||||||||||||||||
Allele Nomenclature: |
|||||||||||||||||||||||||||||||||||||||
Minor Allele Frequency: |
1000Genome | Applied Biosystems® | HapMap | |||
---|---|---|---|---|---|
Global
|
Caucasian - Not Available | CEPH (CEU) - Not Available | |||
EAS
|
African American - Not Available | YRI (Yoruba) - Not Available | |||
SAS
|
Chinese - Not Available | CHB (Han Chinese) - Not Available | |||
AFR
|
Japanese - Not Available | JPT (Japanese) - Not Available | |||
EUR
|
|||||
AMR
|
SIL1 - SIL1 nucleotide exchange factor | ||||||
---|---|---|---|---|---|---|
Transcript Accession | SNP Location | SNP Type | Observed Codons | Observed Amino Acid | Protein ID | |
NM_001037633.1 | 1594 | Missense Mutation | AGC,GGC | S,G 451 | NP_001032722.1 | |
NM_022464.4 | 1594 | Missense Mutation | AGC,GGC | S,G 451 | NP_071909.1 | |
XM_011543570.2 | 1594 | Missense Mutation | AGC,GGC | S,G 461 | XP_011541872.1 |