Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
GTACTTTACAGATAAGTTTTACCAG[A/C]CATTTTTAATCAACATTTCCATTTG
Species: |
Human | ||||||||||||||||||||||||||||||||||||||
dbSNP Submissions: |
NA
|
||||||||||||||||||||||||||||||||||||||
Phenotype: |
MIM: 611027 | ||||||||||||||||||||||||||||||||||||||
Literature Links: |
SHCBP1 PubMed Links | ||||||||||||||||||||||||||||||||||||||
Allele Nomenclature: |
|||||||||||||||||||||||||||||||||||||||
Minor Allele Frequency: |
1000Genome | Applied Biosystems® | HapMap | |||
---|---|---|---|---|---|
Global
|
Caucasian - Not Available | CEPH (CEU) - Not Available | |||
EAS
|
African American - Not Available | YRI (Yoruba) - Not Available | |||
SAS
|
Chinese - Not Available | CHB (Han Chinese) - Not Available | |||
AFR
|
Japanese - Not Available | JPT (Japanese) - Not Available | |||
EUR
|
|||||
AMR
|
SHCBP1 - SHC binding and spindle associated 1 | ||||||
---|---|---|---|---|---|---|
Transcript Accession | SNP Location | SNP Type | Observed Codons | Observed Amino Acid | Protein ID | |
NM_001324318.1 | Intron | NP_001311247.1 | ||||
NM_001324319.1 | Intron | NP_001311248.1 | ||||
NM_024745.4 | Intron | NP_079021.3 | ||||
XM_011523335.2 | Intron | XP_011521637.1 |