Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Species: |
Human | ||||||||||||||||||||||||||||||||
dbSNP Submissions: |
NA
|
||||||||||||||||||||||||||||||||
Phenotype: |
MIM: 605659 | ||||||||||||||||||||||||||||||||
Literature Links: |
CLEC2D PubMed Links | ||||||||||||||||||||||||||||||||
Allele Nomenclature: |
|||||||||||||||||||||||||||||||||
Minor Allele Frequency: |
1000Genome | Applied Biosystems® | HapMap | |||
---|---|---|---|---|---|
Global - Not Available | Caucasian - Not Available | CEPH (CEU)
|
|||
EAS - Not Available | African American - Not Available | YRI (Yoruba)
|
|||
SAS - Not Available | Chinese - Not Available | JPT (Japanese)
|
|||
AFR - Not Available | Japanese - Not Available | CHB (Han Chinese)
|
|||
EUR - Not Available | |||||
AMR - Not Available |
CLEC2D - C-type lectin domain family 2 member D | ||||||
---|---|---|---|---|---|---|
Transcript Accession | SNP Location | SNP Type | Observed Codons | Observed Amino Acid | Protein ID | |
NM_001004419.4 | 94 | Missense Mutation | CTG,GTG | L,V 23 | NP_001004419.1 | |
NM_001197317.2 | 94 | Intron | NP_001184246.1 | |||
NM_001197318.2 | 94 | Missense Mutation | CTG,GTG | L,V 23 | NP_001184247.1 | |
NM_001197319.2 | 94 | Intron | NP_001184248.1 | |||
NM_013269.5 | 94 | Missense Mutation | CTG,GTG | L,V 23 | NP_037401.1 |