Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Species: |
Human | ||||||||||||||||||||||||||||||||||||||||||||||||||
dbSNP Submissions: |
NA
|
||||||||||||||||||||||||||||||||||||||||||||||||||
Phenotype: |
MIM: 616561 | ||||||||||||||||||||||||||||||||||||||||||||||||||
Literature Links: |
MIR1470 PubMed Links | ||||||||||||||||||||||||||||||||||||||||||||||||||
Allele Nomenclature: |
|||||||||||||||||||||||||||||||||||||||||||||||||||
Minor Allele Frequency: |
1000Genome | Applied Biosystems® | HapMap | ||||||
---|---|---|---|---|---|---|---|---|
Global
|
Caucasian - Not Available | CEPH (CEU)
|
||||||
EAS
|
African American - Not Available | YRI (Yoruba)
|
||||||
SAS
|
Chinese - Not Available | JPT (Japanese)
|
||||||
AFR
|
Japanese - Not Available | CHB (Han Chinese)
|
||||||
EUR
|
||||||||
AMR
|
MIR1470 - microRNA 1470 | ||||||
---|---|---|---|---|---|---|
There are no transcripts associated with this gene. |
RASAL3 - RAS protein activator like 3 | ||||||
---|---|---|---|---|---|---|
Transcript Accession | SNP Location | SNP Type | Observed Codons | Observed Amino Acid | Protein ID | |
NM_022904.1 | 2270 | Silent Mutation | NP_075055.1 | |||
XM_011528185.1 | 2270 | Silent Mutation | XP_011526487.1 | |||
XM_011528186.1 | 2270 | Silent Mutation | XP_011526488.1 | |||
XM_011528187.1 | 2270 | Silent Mutation | XP_011526489.1 |
WIZ - widely interspaced zinc finger motifs | ||||||
---|---|---|---|---|---|---|
There are no transcripts associated with this gene. |
Set Membership: |
HapMap |