Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
CCCCAACCCCAAGCTAACAGATGGG[T/C]ACCCTGCTTTCAAAAGACCCCACAT
Species: |
Human | ||||||||||||||||||||||||||||||||||||||
dbSNP Submissions: |
NA
|
||||||||||||||||||||||||||||||||||||||
Phenotype: |
|||||||||||||||||||||||||||||||||||||||
Literature Links: |
LOC101928371 PubMed Links | ||||||||||||||||||||||||||||||||||||||
Allele Nomenclature: |
|||||||||||||||||||||||||||||||||||||||
Minor Allele Frequency: |
1000Genome | Applied Biosystems® | HapMap | |||
---|---|---|---|---|---|
Global
|
Caucasian - Not Available | CEPH (CEU) - Not Available | |||
EAS
|
African American - Not Available | YRI (Yoruba) - Not Available | |||
SAS
|
Chinese - Not Available | CHB (Han Chinese) - Not Available | |||
AFR
|
Japanese - Not Available | JPT (Japanese) - Not Available | |||
EUR
|
|||||
AMR
|
LOC101928371 - uncharacterized LOC101928371 | ||||||
---|---|---|---|---|---|---|
There are no transcripts associated with this gene. |
TEX37 - testis expressed 37 | ||||||
---|---|---|---|---|---|---|
Transcript Accession | SNP Location | SNP Type | Observed Codons | Observed Amino Acid | Protein ID | |
NM_152670.2 | 319 | Missense Mutation | CAC,TAC | H,Y 68 | NP_689883.1 | |
XM_005264188.3 | 319 | Missense Mutation | CAC,TAC | H,Y 68 | XP_005264245.3 | |
XM_011532691.1 | 319 | Missense Mutation | CAC,TAC | H,Y 68 | XP_011530993.1 | |
XM_011532692.1 | 319 | Missense Mutation | CAC,TAC | H,Y 68 | XP_011530994.1 |