Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
GAAAAACCTTTTAAAGAAGAAGAAG[A/G]TATTGAATTTTGACCCTGTCCAAAT
Species: |
Human | ||||||||||||||||||||||||||||||||||||||
dbSNP Submissions: |
NA
|
||||||||||||||||||||||||||||||||||||||
Phenotype: |
|||||||||||||||||||||||||||||||||||||||
Literature Links: |
C14orf39 PubMed Links | ||||||||||||||||||||||||||||||||||||||
Allele Nomenclature: |
|||||||||||||||||||||||||||||||||||||||
Minor Allele Frequency: |
1000Genome | Applied Biosystems® | HapMap | |||
---|---|---|---|---|---|
Global
|
Caucasian - Not Available | CEPH (CEU) - Not Available | |||
EAS
|
African American - Not Available | YRI (Yoruba) - Not Available | |||
SAS
|
Chinese - Not Available | CHB (Han Chinese) - Not Available | |||
AFR
|
Japanese - Not Available | JPT (Japanese) - Not Available | |||
EUR
|
|||||
AMR
|
C14orf39 - chromosome 14 open reading frame 39 | ||||||
---|---|---|---|---|---|---|
Transcript Accession | SNP Location | SNP Type | Observed Codons | Observed Amino Acid | Protein ID | |
NM_174978.2 | 1772 | Missense Mutation | NP_777638.2 | |||
XM_011536690.2 | 1772 | Missense Mutation | XP_011534992.1 | |||
XM_011536703.2 | 1772 | Intron | XP_011535005.1 | |||
XM_011536704.1 | 1772 | Intron | XP_011535006.1 | |||
XM_017021247.1 | 1772 | Missense Mutation | XP_016876736.1 | |||
XM_017021248.1 | 1772 | Missense Mutation | XP_016876737.1 | |||
XM_017021249.1 | 1772 | Missense Mutation | XP_016876738.1 | |||
XM_017021250.1 | 1772 | Missense Mutation | XP_016876739.1 | |||
XM_017021251.1 | 1772 | Missense Mutation | XP_016876740.1 |