Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Species: |
Human | |||||||||||||||||||||||||||||||||||||||||
dbSNP Submissions: |
NA
|
|||||||||||||||||||||||||||||||||||||||||
Phenotype: |
MIM: 604722 MIM: 608052 | |||||||||||||||||||||||||||||||||||||||||
Literature Links: |
SH2D3C PubMed Links | |||||||||||||||||||||||||||||||||||||||||
Allele Nomenclature: |
||||||||||||||||||||||||||||||||||||||||||
Minor Allele Frequency: |
1000Genome | Applied Biosystems® | HapMap | ||||||
---|---|---|---|---|---|---|---|---|
Global
|
Caucasian - Not Available | CEPH (CEU) - Not Available | ||||||
EAS
|
African American - Not Available | YRI (Yoruba)
|
||||||
SAS
|
Chinese - Not Available | CHB (Han Chinese) - Not Available | ||||||
AFR
|
Japanese - Not Available | JPT (Japanese) - Not Available | ||||||
EUR
|
||||||||
AMR
|
SH2D3C - SH2 domain containing 3C | ||||||
---|---|---|---|---|---|---|
There are no transcripts associated with this gene. |
TOR2A - torsin family 2 member A | ||||||
---|---|---|---|---|---|---|
Transcript Accession | SNP Location | SNP Type | Observed Codons | Observed Amino Acid | Protein ID | |
NM_001085347.2 | 837 | Intron | NP_001078816.1 | |||
NM_001134430.2 | 837 | Intron | NP_001127902.1 | |||
NM_001134431.2 | 837 | UTR 3 | NP_001127903.1 | |||
NM_001252018.1 | 837 | Intron | NP_001238947.1 | |||
NM_001252021.1 | 837 | Intron | NP_001238950.1 | |||
NM_001252023.1 | 837 | Intron | NP_001238952.1 | |||
NM_130459.3 | 837 | Missense Mutation | NP_569726.2 | |||
XM_011518554.1 | 837 | Intron | XP_011516856.1 |
TTC16 - tetratricopeptide repeat domain 16 | ||||||
---|---|---|---|---|---|---|
There are no transcripts associated with this gene. |
Set Membership: |
HapMap |