Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
TCATTTTCAACACGTAAGAACATGA[C/T]TTTGTCATCTCTTTATGGGGCACCA
Species: |
Human | ||||||||||||||||||||||||||||||||||||||
dbSNP Submissions: |
NA
|
||||||||||||||||||||||||||||||||||||||
Phenotype: |
MIM: 312610 | ||||||||||||||||||||||||||||||||||||||
Literature Links: |
RPGR PubMed Links | ||||||||||||||||||||||||||||||||||||||
Allele Nomenclature: |
|||||||||||||||||||||||||||||||||||||||
Minor Allele Frequency: |
1000Genome | Applied Biosystems® | HapMap | |||
---|---|---|---|---|---|
Global
|
Caucasian - Not Available | CEPH (CEU) - Not Available | |||
EAS
|
African American - Not Available | YRI (Yoruba) - Not Available | |||
SAS
|
Chinese - Not Available | CHB (Han Chinese) - Not Available | |||
AFR
|
Japanese - Not Available | JPT (Japanese) - Not Available | |||
EUR
|
|||||
AMR
|
RPGR - retinitis pigmentosa GTPase regulator | ||||||
---|---|---|---|---|---|---|
Transcript Accession | SNP Location | SNP Type | Observed Codons | Observed Amino Acid | Protein ID | |
NM_000328.2 | Intron | NP_000319.1 | ||||
NM_001034853.1 | Intron | NP_001030025.1 | ||||
XM_005272633.2 | Intron | XP_005272690.1 | ||||
XM_011543940.2 | Intron | XP_011542242.1 | ||||
XM_017029710.1 | Intron | XP_016885199.1 | ||||
XM_017029711.1 | Intron | XP_016885200.1 | ||||
XM_017029712.1 | Intron | XP_016885201.1 |