Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
TCAGTTCCCTTTGGCTGGAAGGACC[A/G]TGGTTGAGTTTTCTTCGAACTGGGC
Species: |
Human | ||||||||||||||||||||||||||||||||||||||
dbSNP Submissions: |
NA
|
||||||||||||||||||||||||||||||||||||||
Phenotype: |
MIM: 147370 | ||||||||||||||||||||||||||||||||||||||
Literature Links: |
IGF1R PubMed Links | ||||||||||||||||||||||||||||||||||||||
Allele Nomenclature: |
|||||||||||||||||||||||||||||||||||||||
Minor Allele Frequency: |
1000Genome | Applied Biosystems® | HapMap | |||
---|---|---|---|---|---|
Global
|
Caucasian - Not Available | CEPH (CEU) - Not Available | |||
EAS
|
African American - Not Available | YRI (Yoruba) - Not Available | |||
SAS
|
Chinese - Not Available | CHB (Han Chinese) - Not Available | |||
AFR
|
Japanese - Not Available | JPT (Japanese) - Not Available | |||
EUR
|
|||||
AMR
|
IGF1R - insulin like growth factor 1 receptor | ||||||
---|---|---|---|---|---|---|
There are no transcripts associated with this gene. |
PGPEP1L - pyroglutamyl-peptidase I-like | ||||||
---|---|---|---|---|---|---|
Transcript Accession | SNP Location | SNP Type | Observed Codons | Observed Amino Acid | Protein ID | |
NM_001102612.2 | 867 | Missense Mutation | ACG,ATG | T,M 189 | NP_001096082.2 | |
NM_001167902.1 | 867 | Intron | NP_001161374.1 | |||
XM_011521254.1 | 867 | Missense Mutation | ACG,ATG | T,M 280 | XP_011519556.1 | |
XM_017021933.1 | 867 | Missense Mutation | ACG,ATG | T,M 135 | XP_016877422.1 |