Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
TGACAGATCTGGGAGGCCTTGACCC[T/C]GTGTGGCTGTTTGTGGTAGTTGGAG
Species: |
Human | ||||||||||||||||||||||||||||||||||||||
dbSNP Submissions: |
NA
|
||||||||||||||||||||||||||||||||||||||
Phenotype: |
|||||||||||||||||||||||||||||||||||||||
Literature Links: |
EIF4E1B PubMed Links | ||||||||||||||||||||||||||||||||||||||
Allele Nomenclature: |
|||||||||||||||||||||||||||||||||||||||
Minor Allele Frequency: |
1000Genome | Applied Biosystems® | HapMap | |||
---|---|---|---|---|---|
Global
|
Caucasian - Not Available | CEPH (CEU) - Not Available | |||
EAS
|
African American - Not Available | YRI (Yoruba) - Not Available | |||
SAS
|
Chinese - Not Available | CHB (Han Chinese) - Not Available | |||
AFR
|
Japanese - Not Available | JPT (Japanese) - Not Available | |||
EUR
|
|||||
AMR
|
EIF4E1B - eukaryotic translation initiation factor 4E family member 1B | ||||||
---|---|---|---|---|---|---|
There are no transcripts associated with this gene. |
TSPAN17 - tetraspanin 17 | ||||||
---|---|---|---|---|---|---|
Transcript Accession | SNP Location | SNP Type | Observed Codons | Observed Amino Acid | Protein ID | |
NM_001006616.2 | 418 | Silent Mutation | CCC,CCT | P,P 63 | NP_001006617.2 | |
NM_012171.2 | 418 | Silent Mutation | CCC,CCT | P,P 63 | NP_036303.1 | |
NM_130465.4 | 418 | Silent Mutation | CCC,CCT | P,P 63 | NP_569732.2 | |
XM_005265876.3 | 418 | Intron | XP_005265933.1 |